missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-butanol, 97%

Product Code. 10315403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10315403 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10315403 Supplier Thermo Scientific Acros Supplier No. 183400050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2344-70-9 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2344-70-9
Density 0.9800g/mL
Boiling Point 132.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O
Linear Formula C6H5CH2CH2CH(OH)CH3
Refractive Index 1.5157 to 1.5177
Quantity 5 g
Beilstein 06,522
Specific Gravity 0.98
Synonym 4-phenyl-2-butanol, 1-phenyl-3-butanol, 2-hydroxy-4-phenylbutane, methyl phenethyl carbinol, methylphenethylcarbinol, 2-butanol, 4-phenyl, alpha-methylbenzenepropanol, phenylethyl methyl carbinol, methyl-2-phenylethylcarbinol
InChI Key GDWRKZLROIFUML-UHFFFAOYSA-N
SMILES CC(CCC1=CC=CC=C1)O
IUPAC Name 4-phenylbutan-2-ol
Molecular Weight (g/mol) 150.22
PubChem CID 61302
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.