missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-butanone, 98%

Product Code. 11439817
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439817 250 g 250g
11429817 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439817 Supplier Thermo Scientific Alfa Aesar Supplier No. B22777.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2550-26-7 | C10H12O | 148.205 g/mol

4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-26-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00008790
InChI Key AKGGYBADQZYZPD-UHFFFAOYSA-N
Synonym benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
PubChem CID 17355
IUPAC Name 4-phenylbutan-2-one
SMILES CC(=O)CCC1=CC=CC=C1

Specifications

Density 0.988
Boiling Point 235°C to 237°C
Flash Point 105°C (221°F)
Refractive Index 1.5122
Quantity 250 g
Beilstein 1907123
Solubility Information Not miscible in water.
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-Phenyl-2-butanone
compliance-icons
Product Identifier
  • 4-Phenyl-2-butanone
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-phenyl-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.