missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-ol, 97%

Product Code. 15410278
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410278 5 g 5g
15420278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410278 Supplier Thermo Scientific Alfa Aesar Supplier No. H53450.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5876-76-6 | C10H10O | 146.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5876-76-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00046651
InChI Key JYOZFNMFSVAZAW-UHFFFAOYSA-N
Synonym 4-phenyl-3-butyn-2-ol, 4-phenyl-3-butyne-2-ol, 3-hydroxy-1-phenyl-1-butyne
PubChem CID 582962
IUPAC Name 4-phenylbut-3-yn-2-ol
SMILES CC(C#CC1=CC=CC=C1)O

Specifications

Density 1.005
Flash Point >110°C (230°F)
Refractive Index 1.5667
Quantity 5 g
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 4-Phenyl-3-butyn-2-ol
compliance-icons
Product Identifier
  • 4-Phenyl-3-butyn-2-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Phenyl-3-butyn-2-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.