missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-one, 98%

Product Code. 15373148 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15373148 5 g 5g
15343148 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15373148 Supplier Thermo Scientific Acros Supplier No. 171330050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1817-57-8 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-57-8
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
InChI Key UPEUQDJSUFHFQP-UHFFFAOYSA-N
Synonym 4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
PubChem CID 74555
ChEBI CHEBI:51731
IUPAC Name 4-phenylbut-3-yn-2-one
SMILES CC(=O)C#CC1=CC=CC=C1

Specifications

Density 0.9900g/mL
Boiling Point 75°C to 76°C (1.1 hPa)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5C≡CCOCH3
Refractive Index 1.5762
Quantity 5 g
Beilstein 07,385
Specific Gravity 0.99
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material 4-Phenyl-3-butyn-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenyl-3-butyn-2-one, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.