missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-5-(trifluoromethyl)thiophen-3-amine, ≥97%, Thermo Scientific™

Product Code. 10343153
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10343153 250 mg 250mg
10231043 1 g 1g
10832051 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10343153 Supplier Thermo Scientific Maybridge Supplier No. SEW03133CB
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 256427-77-7
Molecular Formula C11H8F3NS
Molecular Weight (g/mol) 243.247
MDL Number MFCD01934528
InChI Key KBIPYEXWOYDDDL-UHFFFAOYSA-N
Synonym 4-phenyl-5-trifluoromethyl thiophen-3-amine, 4-phenyl-5-trifluoromethyl thien-3-ylamine, 3-amino-4-phenyl-5-trifluoromethyl thiophene, maybridge3_006438, 4-phenyl-5-trifluoromethyl-3-thienylamine, 4-phenyl-5-trifluoromethyl thiophene-3-amine
PubChem CID 2782905
IUPAC Name 4-phenyl-5-(trifluoromethyl)thiophen-3-amine
SMILES C1=CC=C(C=C1)C2=C(SC=C2N)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 243.25
Percent Purity 97%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophen-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.