missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10149602
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10149602 250 mg Amber Glass Bottle 250mg
10644603 1 g Amber Glass Bottle 1g
10215334 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10149602 Supplier Thermo Scientific Maybridge Supplier No. SEW02660CB
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 208108-76-3
Molecular Formula C12H7F3O2S
Molecular Weight (g/mol) 272.241
InChI Key GIUAGKGTDDIMHB-UHFFFAOYSA-N
Synonym 4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid
PubChem CID 2783372
IUPAC Name 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 272.25
Percent Purity 95%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.