missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 97%

Product Code. 10722602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722602 25 g Glass bottle 25g
10589803 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722602 Supplier Thermo Scientific Acros Supplier No. 417360250
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1689-82-3 | C12H10N2O | 198.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Beilstein 16, 96
Color Brown
Quantity 25 g
Physical Form Fine Crystalline Powder
Formula Weight 198.22
Percent Purity ≥96.0%
Chemical Name or Material 4-Phenylazophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenylazophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.