missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, TRC

Product Code. 30428002
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428002 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428002 Supplier Toronto Research Chemicals Supplier No. TRCP319640100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Phenylazophenol
CAS 1689-82-3
Molecular Formula C12H10N2O
Quantity 100 g
Synonym 4-(Phenyldiazenyl)phenol, 4-Hydroxyazobenzene, Brasilazina Oil Yellow O, C.I. 11800, NSC 3177, Pirocard Green 491, Solvent Yellow 7, p-(Phenylazo)phenol, p-Hydroxyazobenzene, C.I. Solvent Yellow 7, 4-(Phenylazo)phenol
InChI Formula InChI=1S/C12H10N2O/c15-12-8-6-11(7-9-12)14-13-10-4-2-1-3-5-10/h1-9,15H/b14-13+
SMILES Oc1ccc(cc1)N=Nc2ccccc2
IUPAC Name 4-[(E)-phenyldiazenyl]phenol
Molecular Weight (g/mol) 198.22
Formula Weight 198.0793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.