missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, 99%

Product Code. 11439276
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11439276 25 g 25g
11449276 100 g 100g
11459276 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11439276 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A18115.14
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1821-12-1 | C10H12O2 | 164.204 g/mol

4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

Solubility
Soluble in water and methanol.

Notes
Styrene-DVB; Strongly basic gel type resin.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1821-12-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00004403
InChI Key OBKXEAXTFZPCHS-UHFFFAOYSA-N
Synonym 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
PubChem CID 4775
ChEBI CHEBI:41500
IUPAC Name 4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCCC(=O)O

Spécification

Melting Point 49°C to 53°C
Boiling Point 165°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 638180
Solubility Information Soluble in water and methanol.
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 4-Phenylbutyric acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.