missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, 99%

Product Code. 11954981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11954981 500 g Glass bottle 500g
10614802 25 g Glass bottle 25g
10583291 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11954981 Supplier Thermo Scientific Acros Supplier No. 130385000
 Restricted Item
Estimated Shipment: 02-06-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1821-12-1 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1821-12-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
InChI Key OBKXEAXTFZPCHS-UHFFFAOYSA-N
Synonym 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
PubChem CID 4775
ChEBI CHEBI:41500
IUPAC Name 4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 49.0°C to 52.0°C
Color White to Yellow
Boiling Point 165.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5(CH2)3CO2H
Quantity 500 g
Beilstein 09,539
Solubility Information Solubility in water: 5.3g/L water (40 degr. c)
Formula Weight 164.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.