missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric Acid, TRC

Product Code. 30416017
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416017 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416017 Supplier Toronto Research Chemicals Supplier No. TRCP3196102.5G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1821-12-1
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym Benzenebutanoic acid, Butyric acid, 4-phenyl- (8CI), Butyric acid, γ-phenyl- (3CI), 4-Phenyl-n-butyric acid, 4-Phenylbutanoic acid, 4-Phenylbutyric acid, Benzenebutyric acid, NSC 295, γ-Phenylbutanoic acid, γ-Phenylbutyric acid, ω-Phenylbutanoic acid, Phenylbutyric acid
IUPAC Name 4-phenylbutanoic acid
SMILES OC(=O)CCCc1ccccc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)
Formula Weight 164.0837
Chemical Name or Material 4-Phenylbutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.