missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyryl chloride, 97%

Product Code. 11345379
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345379 5 g 5g
11335379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345379 Supplier Thermo Scientific Alfa Aesar Supplier No. H50388.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18496-54-3 | C10H11ClO | 182.647 g/mol

4-Phenylbutyryl chloride is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylbutyryl chloride is used as organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, active metals, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18496-54-3
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00798080
InChI Key VQDQISMDUHBUFF-UHFFFAOYSA-N
Synonym 4-phenylbutyryl chloride, 4-phenylbutanoylchloride, 4-phenyl-butyryl chloride, 4-phenylbutyric acid chloride, acmc-1cuil, 4-phenyl butyryl chloride, 4-phenyl-mutanoyl chloride, 4-phenyl-butyric acid chloride
PubChem CID 11789865
IUPAC Name 4-phenylbutanoyl chloride
SMILES C1=CC=C(C=C1)CCCC(=O)Cl

Especificaciones

Boiling Point 273°C
Linear Formula C6H5CH2CH2CH2COCl
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material 4-Phenylbutyryl chloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.