missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine, 98%

Product Code. 10673362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10673362 1 g Glass bottle 1g
10371464 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10673362 Supplier Thermo Scientific Acros Supplier No. 164770010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 771-99-3 | C11H15N | 161.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-99-3
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.25
MDL Number MFCD00006002
InChI Key UTBULQCHEUWJNV-UHFFFAOYSA-N
Synonym 4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
PubChem CID 69873
IUPAC Name 4-phenylpiperidine
SMILES C1CNCCC1C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 57.0°C to 63.0°C
Color White to Brown
Boiling Point 255.0°C to 257.0°C (727.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 291
Formula Weight 161.25
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Phenylpiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.