missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylurazole, 98+%

Product Code. 11339963
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339963 25 g 25g
11329963 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339963 Supplier Thermo Scientific Alfa Aesar Supplier No. L00255.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15988-11-1 | C8H7N3O2 | 177.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15988-11-1
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00005226
InChI Key GOSUFRDROXZXLN-UHFFFAOYSA-N
Synonym 4-phenylurazole, urazole, 4-phenyl, 1,2,4-triazolidine-3,5-dione, 4-phenyl, bicarbamimide, n-phenyl, 4-phenyl urazole, 4-phenylurazol, 4-phenyl-urazole, acmc-1c52y, ksc297q5n
PubChem CID 85229
IUPAC Name 4-phenyl-1,2,4-triazolidine-3,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)NNC2=O

Specifications

Melting Point 206°C to 210°C
Quantity 25 g
Beilstein 163361
Formula Weight 177.16
Percent Purity ≥98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 4-Phenylurazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.