missing translation for 'onlineSavingsMsg'
Learn More

4-Phthalimidobutyric Acid, TRC

Product Code. 30376896
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376896 250 mg -
30371305 500 mg 500mg
30388132 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376896 Supplier Toronto Research Chemicals Supplier No. TRCD486408250MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3130-75-4
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.22
Synonym 4-Phthalimidoylbutyric Acid, N-Phthalyl-gamma-aminobutyric Acid, NSC 119133, Phthaloyl gamma-Aminobutyric Acid, gamma-Phthalimidobutyric Acid, 1,3-Dihydro-1,3-dioxo-2H-isoindole-2-butanoic Acid, 4-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)butanoic Acid, 4-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)butyric Acid, 4-(1,3-Dioxoisoindolin-2-yl)butanoic Acid, 4-Phthalimidobutanoic Acid
SMILES OC(=O)CCCN1C(=O)c2ccccc2C1=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H11NO4/c14-10(15)6-3-7-13-11(16)8-4-1-2-5-9(8)12(13)17/h1-2,4-5H,3,6-7H2,(H,14,15)
Formula Weight 233.0688
Chemical Name or Material 4-Phthalimidobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.