missing translation for 'onlineSavingsMsg'
Learn More

4-(Piperazin-1-yl)thieno[3,2-c]pyridine Dihydrochloride, TRC

Product Code. 30424370
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424370 10 mg 10mg
30414013 50 mg -
30408858 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424370 Supplier Toronto Research Chemicals Supplier No. TRCP48254310MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2514947-85-2
Molecular Formula C11H13N3S . 2HCl
Molecular Weight (g/mol) 219.31
IUPAC Name 4-piperazin-1-ylthieno[3,2-c]pyridine
SMILES C1CN(CCN1)c2nccc3sccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H13N3S/c1-3-13-11(9-2-8-15-10(1)9)14-6-4-12-5-7-14/h1-3,8,12H,4-7H2
Formula Weight 219.083
Chemical Name or Material 4-(Piperazin-1-yl)thieno[3,2-c]pyridine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.