missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidin-1-ylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10472654
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10472654 1 g Amber Glass Bottle 1g
10622515 10 g Amber Glass Bottle 10g
10245764 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10472654 Supplier Thermo Scientific Maybridge Supplier No. CC33916DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1204-85-9
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD02575368
InChI Key ZEPXHFFGXQFUDP-UHFFFAOYSA-N
Synonym 4-piperidin-1-yl benzonitrile, 4-1-piperidinyl benzonitrile, 4-piperidin-1-yl-benzonitrile, 4-piperidinobenzonitrile, benzonitrile,4-1-piperidinyl, 4-piperidylbenzenecarbonitrile, acmc-209a7n, 4-piperidin-1-ylbenzenecarbonitrile
PubChem CID 766771
IUPAC Name 4-piperidin-1-ylbenzonitrile
SMILES C1CCN(CC1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.26
Percent Purity 97%
Chemical Name or Material 4-Piperidin-1-ylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.