missing translation for 'onlineSavingsMsg'
Learn More

[4-(Piperidin-1-ylmethyl)phenyl]methanol, ≥95%, Thermo Scientific™

Product Code. 10609564
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10609564 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10609564 Supplier Thermo Scientific Maybridge Supplier No. CC45509DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 91271-62-4
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H19NO
MDL Number MFCD07772863
Quantity 1 g
Synonym 4-piperidin-1-ylmethyl phenyl methanol, 4-piperidylmethyl phenyl methan-1-ol, benzenemethanol,4-1-piperidinylmethyl, 4-piperidin-1-yl methyl benzyl alcohol, benzenemethanol, 4-1-piperidinylmethyl, 4-piperidin-1-yl methyl phenyl methanol
InChI Key SQWKPHFBCVNPGU-UHFFFAOYSA-N
SMILES C1CCN(CC1)CC2=CC=C(C=C2)CO
IUPAC Name [4-(piperidin-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol) 205.301
PubChem CID 7162041
Formula Weight 205.3
Percent Purity 95%
Chemical Name or Material [4-(Piperidin-1-ylmethyl)phenyl]methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.