missing translation for 'onlineSavingsMsg'
Learn More

4-(Piperidinosulfonyl)aniline, 97%, Thermo Scientific™

Product Code. 10541513
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10541513 10 g 10g
10305402 1 g 1g
10361815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10541513 Supplier Thermo Scientific Maybridge Supplier No. TL00817EA
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6336-68-1
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD00276969
InChI Key ZTTBIWZAAMPNBE-UHFFFAOYSA-N
Synonym 4-piperidin-1-ylsulfonyl aniline, 4-1-piperidinylsulfonyl aniline, 4-piperidine-1-sulfonyl aniline, 4-piperidinosulfonyl aniline, 4-piperidine-1-sulfonyl-phenylamine, 4-piperidylsulfonyl phenylamine, maybridge1_000153, cambridge id 5358277, acmc-1bh47, p-piperidinylsulfonyl phenylamine
PubChem CID 236648
IUPAC Name 4-piperidin-1-ylsulfonylaniline
SMILES NC1=CC=C(C=C1)S(=O)(=O)N1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 240.33
Percent Purity 97%
Chemical Name or Material 4-(Piperidinosulfonyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.