missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-17alpha,21-diol-3,20-dione-21,21-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 11-Deoxycortisol-21,21-D2 |
| CAS | 1271728-08-5 |
| Molecular Formula | C21 D2 H28 O4 |
| Quantity | 0.01 g |
| Synonym | Pregn-4-ene-3,20-dione-21,21-d2, 17,21-dihydroxy- (ACI), 17,21-Dihydroxypregn-4-ene-3,20-dione-21,21-d2 (ACI), 11-Deoxycortisol-21,21-d2, 11-Deoxycortisol-21,21-D2, 4-Pregnen-17α,21-diol-3,20-dione-21,21-d2 |
| InChI Formula | InChI=1 S/C21H30O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h11,15-17,22,25 H,3-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1/i12D2 |
| SMILES | [2 H]C([2 H])(O)C(=O)[C@@]1(O)CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 R)-17-(2,2-dideuterio-2-hydroxyacetyl)-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 348.4728 |
| Formula Weight | 348.227 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction