missing translation for 'onlineSavingsMsg'
Learn More

4-Prop-2-ynyl-thiomorpholine 1,1-dioxide, 97%, Thermo Scientific™

Product Code. 10758764
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10758764 1 g Amber Glass Bottle 1g
10391663 250 mg Amber Glass Bottle 250mg
10603055 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10758764 Supplier Thermo Scientific Maybridge Supplier No. DFP00032DA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10442-03-2
Molecular Formula C7H11NO2S
Molecular Weight (g/mol) 173.23
InChI Key CZBZIZOYSYHBNM-UHFFFAOYSA-N
PubChem CID 2777265
IUPAC Name 4-prop-2-ynyl-1,4-thiazinane 1,1-dioxide
SMILES C#CCN1CCS(=O)(=O)CC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-propargylthiomorpholine 1,1-dioxide, 4-prop-2-yn-1-yl thiomorpholine 1,1-dioxide, 4-2-propynyl thiomorpholine 1,1-dioxide, 4-propargyl thiomorpholine-1,1-dioxide, 4-propargylthiomorpholine1,1-dioxide, thiomorpholine, 4-2-propyn-1-yl-, 1,1-dioxide, 4-prop-2-yn-1-yl-1??-thiomorpholine-1,1-dione, acmc-1byom, 4-propargylthiomorpholine-1,1-dioxide, 4-propargyl-thiomorpholine-1,1-dioxide
Formula Weight 173.23
Percent Purity 97%
Chemical Name or Material 4-(Prop-2-ynyl)thiomorpholine 1, 1-dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.