missing translation for 'onlineSavingsMsg'
Learn More

4-(Propargylaminocarbonyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15439175
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439175 5 g 5g
15429175 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439175 Supplier Thermo Scientific Alfa Aesar Supplier No. H51125.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-49-8
Molecular Formula C16H20BNO3
Molecular Weight (g/mol) 285.15
MDL Number MFCD11113041
InChI Key BUZIZNWTLHGQQO-UHFFFAOYSA-N
Synonym 4-2-propynylcarbamoyl phenylboronic acid pinacol ester, n-prop-2-yn-1-yl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-propargylaminocarbonyl phenylboronic acid pinacol ester, n-prop-2-yn-1-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-prop-2-ynylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb136, 4-propargylaminocarbonyl benzeneboronic acid pinacol ester, n-2-propynyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-propynyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl benzamide
PubChem CID 46739749
IUPAC Name N-prop-2-ynyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCC#C

Specifications

Quantity 5 g
Formula Weight 285.15
Percent Purity 95%
Chemical Name or Material 4-(Propargylaminocarbonyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.