missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrazoleboronic Acid Pinacol Ester, TRC

Product Code. 30423310
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423310 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423310 Supplier Toronto Research Chemicals Supplier No. TRCP8422002.5G
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pyrazoleboronic Acid Pinacol Ester
CAS 269410-08-4
Molecular Formula C9H15BN2O2
Quantity 2.5 g
Synonym (1H-Pyrazol-4-yl)boronic acid pinacol ester, 1H-Pyrazole-4-boronic Acid Pinacol Ester, 2-(1H-Pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(1H-pyrazol-2-yl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(1H-pyrazol-4-yl)-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, Pyrazol-4-ylboronic Acid Pinacol Ester
InChI Formula InChI=1S/C9H15BN2O2/c1-8(2)9(3,4)14-10(13-8)7-5-11-12-6-7/h5-6H,1-4H3,(H,11,12)
SMILES CC1(C)OB(OC1(C)C)c2cn[nH]c2
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Weight (g/mol) 194.04
Formula Weight 194.1227
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.