missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrid-4-ylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10036212
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10036212 1 g Amber Glass Bottle 1g
10004332 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10036212 Supplier Thermo Scientific Maybridge Supplier No. CC35401DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4385-76-6
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD04114574
InChI Key DZLGZIGLHCRIMF-UHFFFAOYSA-N
Synonym 4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv
PubChem CID 1520811
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-(pyridin-4-yl)benzoic acid
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 4-Pyrid-4-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.