missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridazinecarboxylic acid, 95%

Product Code. 10143044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10143044 1 g 1g
10284614 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10143044

Brand: Thermo Scientific Acros 399630010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50681-25-9 | C5H4N2O2 | 124.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50681-25-9
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
InChI Key JUSIWJONLKBPDU-UHFFFAOYSA-N
Synonym 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid
PubChem CID 2761046
IUPAC Name pyridazine-4-carboxylic acid
SMILES C1=CN=NC=C1C(=O)O

Specifications

Melting Point 244.2°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 124.1
Percent Purity 95%
Chemical Name or Material 4-Pyridazinecarboxylic acid
compliance-icons
Product Identifier
  • 4-Pyridazinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.