missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetonitrile, ≥97%, Thermo Scientific Chemicals

Product Code. 15431869
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431869 1 g 1g
15441869 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431869 Supplier Thermo Scientific Alfa Aesar Supplier No. H64588.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 13121-99-8 | Molecular Formula: C7H6N2 | Molecular Weight: 118.14 g/mol

A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13121-99-8
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD03412601
InChI Key BMVSAKPRNWZCPG-UHFFFAOYSA-N
Synonym pyridin-4-yl-acetonitrile, 2-pyridin-4-yl acetonitrile, 4-pyridylacetonitrile, 4-pyridineacetonitrile, pyridin-4-ylacetonitrile, pyridine-4-acetonitrile, rarechem ak ml 0204, 2-4-pyridyl acetonitrile, 4-pyridylacetonitrilehydrochloride, 4-cyanomethyl-pyridine
PubChem CID 4112085
IUPAC Name 2-pyridin-4-ylacetonitrile
SMILES N#CCC1=CC=NC=C1

Specifica

Quantity 1 g
UN Number UN3439
Beilstein 471559
Solubility Information Slightly soluble in water.
Formula Weight 118.14
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 4-Pyridineacetonitrile

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.