missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineboronic Acid, TRC

Product Code. 30413486
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413486 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413486 Supplier Toronto Research Chemicals Supplier No. TRCP99135525G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pyridineboronic Acid
CAS 1692-15-5
Molecular Formula C5 H6 B N O2
Quantity 25 g
InChI Formula InChI=1S/C5H6BNO2/c8-6(9)5-1-3-7-4-2-5/h1-4,8-9H
SMILES OB(O)c1ccncc1
IUPAC Name pyridin-4-ylboronic acid
Molecular Weight (g/mol) 122.92
Formula Weight 123.0492
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.