missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinecarboxaldehyde, TRC

Product Code. 30413444
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30413444 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30413444

Brand: Toronto Research Chemicals TRCP991455100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pyridinecarboxaldehyde
CAS 872-85-5
Molecular Formula C6H5NO
Quantity 100 g
Synonym Isonicotinaldehyde, 4-Formylpyridine, 4-Pyridinealdehyde, 4-Pyridylcarboxyaldehyde, Isonicotinic Aldehyde, NSC 8953, Pyridin-4-al, Pyridin-4-carboxaldehyde, Pyridin-4-ylcarboxaldehyde, Pyridine-4-carbaldehyde, p-Formylpyridine, p-Pyridylaldehyde, gamma-Formylpyridine
InChI Formula InChI=1S/C6H5NO/c8-5-6-1-3-7-4-2-6/h1-5H
SMILES O=Cc1ccncc1
IUPAC Name pyridine-4-carbaldehyde
Molecular Weight (g/mol) 107.11
Formula Weight 107.0371
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.