missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinepropanol, Thermo Scientific Chemicals

Product Code. 17261152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17261152 25 g 25g
17251152 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17261152 Supplier Thermo Scientific Acros Supplier No. 440560250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2629-72-3 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2629-72-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key PZVZGDBCMQBRMA-UHFFFAOYSA-N
IUPAC Name 3-(pyridin-4-yl)propan-1-ol
SMILES OCCCC1=CC=NC=C1

Specifications

Melting Point 35.0°C to 39.0°C
Boiling Point 289.0°C
Flash Point 113°C
MDL Number MFCD00047457
Quantity 25 g
Formula Weight 137.81
Chemical Name or Material 4-Pyridinepropanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.