missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridylacetic Acid Hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10713071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10713071 5 g 5g
10057732 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713071 Supplier Thermo Scientific Acros Supplier No. 294750050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6622-91-9 | Molecular Formula: C7H7NO2·HCl | Molecular Weight: 173.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-91-9
MDL Number MFCD00012827
InChI Key WKJRYVOTVRPAFN-UHFFFAOYSA-N
Synonym 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride
PubChem CID 81097
IUPAC Name 2-pyridin-4-ylacetic acid;hydrochloride
SMILES C1=CN=CC=C1CC(=O)O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7H8ClNO2
Linear Formula (C5H4N)CH2COOH.HCl
Quantity 5 g
Molecular Weight (g/mol) 173.596
Formula Weight 173.6
Percent Purity ≥97.5%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Pyridylacetic acid hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Pyridylacetic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.