missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylaniline, 95%, Thermo Scientific™

Product Code. 10682855
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10682855 1 g Amber Glass Bottle 1g
10245654 250 mg Amber Glass Bottle 250mg
2 options
This item is not returnable. View return policy

Product Code. 10682855

Brand: Thermo Scientific Maybridge CC56214DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69491-57-2
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.203
MDL Number MFCD09817490
InChI Key RUVNVJCKWOOLOH-UHFFFAOYSA-N
PubChem CID 12456170
IUPAC Name 4-pyrimidin-2-ylaniline
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-pyrimidin-2-yl aniline, 2-4-aminophenyl pyrimidine, p-aminophenylpyrimidine, 4-2-pyrimidinyl aniline, 4-pyrimidin-2-ylphenylamine, benzenamine,4-2-pyrimidinyl, benzenamine, 4-2-pyrimidinyl
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material 4-Pyrimidin-2-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.