missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzonitrile, 95%, Thermo Scientific™

Product Code. 10692855
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10692855 5 g Amber Glass Bottle 5g
10217153 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10692855

Brand: Thermo Scientific Maybridge CC56216DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78322-96-0
Molecular Formula C11H7N3
Molecular Weight (g/mol) 181.198
MDL Number MFCD09817491
InChI Key WGCKVXHOTDDNOA-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzonitrile, 4-cyanophenyl pyrimidine, 2-4-cyanophenyl pyrimidine, 4-pyrimidin-2-yl-benzonitrile, 4-2-pyrimidinyl benzonitrile, benzonitrile, 4-2-pyrimidinyl, 4-pyrimidin-2-ylbenzenecarbonitrile
PubChem CID 15486310
IUPAC Name 4-pyrimidin-2-ylbenzonitrile
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 181.2
Percent Purity 95%
Chemical Name or Material 4-Pyrimidin-2-ylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.