missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrrolidinopyridine, TRC

Product Code. 30423564
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423564 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423564 Supplier Toronto Research Chemicals Supplier No. TRCP99757025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pyrrolidinopyridine
CAS 2456-81-7
Molecular Formula C9H12N2
Quantity 25 g
Synonym 4-(1-Pyrrolidinyl)-pyridine, 4-(1-Pyrrolidino)pyridine, 4-(1-Pyrrolidinyl)pyridine, 4-(N-Pyrrolidinyl)pyridine
InChI Formula InChI=1S/C9H12N2/c1-2-8-11(7-1)9-3-5-10-6-4-9/h3-6H,1-2,7-8H2
SMILES C1CCN(C1)c2ccncc2
IUPAC Name 4-pyrrolidin-1-ylpyridine
Molecular Weight (g/mol) 148.2
Formula Weight 148.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.