missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfamoylbenzoic Acid, TRC

Product Code. 30416081
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416081 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416081 Supplier Toronto Research Chemicals Supplier No. TRCS6993455G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Sulfamoylbenzoic Acid
CAS 138-41-0
Molecular Formula C7H7NO4S
Quantity 5 g
Synonym 4-(Aminosulfonyl)benzoic Acid, 4-Carboxybenzenesulfonamide, 4-Sulfamoylbenzoic Acid, Benzoic Acid p-Sulfamide, Carzenide, Dirnate, NSC 22976, NSC 683540, p-Aminobenzoic Acid Sulfamide, p-Aminosulfonylbenzoic Acid, p-Carboxybenzenesulfonamide, p-Sulfamoylbenzoic Acid, p-Sulfamylbenzoic Acidp-Sulfonamidobenzoic Acid
InChI Formula InChI=1S/C7H7NO4S/c8-13(11,12)6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)(H2,8,11,12)
SMILES NS(=O)(=O)c1ccc(cc1)C(=O)O
IUPAC Name 4-sulfamoylbenzoic acid
Molecular Weight (g/mol) 201.2
Formula Weight 201.0096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.