missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfophthalic Acid (50% w/w aq. soln.) (may contain up to 20% 3-Sulfophthalic Acid), TRC
Specifications
Specifications
| Chemical Name or Material | 4-Sulfophthalic Acid |
| CAS | 89-08-7 |
| Molecular Formula | C8H6O7S |
| Quantity | 100 mL |
| Synonym | 4-Sulfo-1,2-benzenedicarboxylic Acid, 4-Sulfo-phthalic Acid, NSC 100615, NSC 163994 |
| InChI Formula | InChI=1S/C8H6O7S/c9-7(10)5-2-1-4(16(13,14)15)3-6(5)8(11)12/h1-3H,(H,9,10)(H,11,12)(H,13,14,15) |
| SMILES | OC(=O)c1ccc(cc1C(=O)O)S(=O)(=O)O |
| IUPAC Name | 4-sulfophthalic acid |
| Molecular Weight (g/mol) | 246.19 |
| Formula Weight | 245.9834 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?