missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfophthalic Acid (50% w/w aq. soln.) (may contain up to 20% 3-Sulfophthalic Acid), TRC

Product Code. 30413701
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413701 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413701 Supplier Toronto Research Chemicals Supplier No. TRCS890870250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Sulfophthalic Acid
CAS 89-08-7
Molecular Formula C8H6O7S
Quantity 250 mL
Synonym 4-Sulfo-1,2-benzenedicarboxylic Acid, 4-Sulfo-phthalic Acid, NSC 100615, NSC 163994
InChI Formula InChI=1S/C8H6O7S/c9-7(10)5-2-1-4(16(13,14)15)3-6(5)8(11)12/h1-3H,(H,9,10)(H,11,12)(H,13,14,15)
SMILES OC(=O)c1ccc(cc1C(=O)O)S(=O)(=O)O
IUPAC Name 4-sulfophthalic acid
Molecular Weight (g/mol) 246.19
Formula Weight 245.9834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.