missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15439957
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439957 1 g 1g
15420476 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439957 Supplier Thermo Scientific Alfa Aesar Supplier No. H53198.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-54-6
Molecular Formula C11H15BO4
Molecular Weight (g/mol) 222.047
MDL Number MFCD03411946
InChI Key QMVMDYSTJSUDKC-UHFFFAOYSA-N
Synonym 4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid
PubChem CID 2773301
IUPAC Name [4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O

Specifications

Melting Point 100°C to 106°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 222.05
Percent Purity 95%
Chemical Name or Material 4-(tert-Butoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.