missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Product Code. 10663455
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10663455 10 g 10g
10275284 1 g 1g
10188494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663455 Supplier Thermo Scientific Maybridge Supplier No. CD08308EA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 74370-93-7
Molecular Formula C7H12N2S
Molecular Weight (g/mol) 156.247
MDL Number MFCD00051644
InChI Key CUWZBHVYLVGOAB-UHFFFAOYSA-N
Synonym 2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
PubChem CID 2734202
IUPAC Name 4-tert-butyl-1,3-thiazol-2-amine
SMILES CC(C)(C)C1=CSC(=N1)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 156.25
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)-1, 3-thiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.