missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals

Product Code. 16617022 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 16617022

Brand: Thermo Scientific Acros 464350050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947725-04-4
Molecular Formula C12H17F3S
Molecular Weight (g/mol) 250.32
MDL Number MFCD11858620
InChI Key VRTQPEYVMHATOA-UHFFFAOYSA-N
IUPAC Name 5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
SMILES CC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C

Specifications

Alpha Vector BUTYLDIMETHYLPHENYLSULFURTRIFLUORIDE
Melting Point 62°C to 68°C
Color Red to White
Boiling Point 92°C to 93°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
UN Number 2923
Sensitivity Moisture sensitive
Solubility Information insoluble aq. soln., soluble in hexane, acetonitrile, dichloromethane
Formula Weight 250.32
Physical Form Crystals or Powder
Chemical Name or Material 4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.