missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Alpha Vector | BUTYLDIMETHYLPHENYLSULFURTRIFLUORIDE |
| Melting Point | 62°C to 68°C |
| Color | Red to White |
| Boiling Point | 92°C to 93°C |
| Infrared Spectrum | Conforms |
| Packaging | glass bottle |
| Quantity | 25 g |
| UN Number | 2923 |
| Sensitivity | Moisture sensitive |
| Solubility Information | insoluble aq. soln., soluble in hexane, acetonitrile, dichloromethane |
| Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu