missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl-d9-phenol-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-tert-Butyl-d9-phenol-2,3,5,6-d4 |
| CAS | 225386-58-3 |
| Molecular Formula | C10 D13 H O |
| Quantity | 0.25 g |
| Synonym | Phen-2,3,5,6-d4-ol, 4-[1,1-di(methyl-d3)ethyl-2,2,2-d3]- (9 CI) |
| InChI Formula | InChI=1 S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 163.3 |
| Formula Weight | 163.1861 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction