missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)piperidine hydrochloride, 90%, Thermo Scientific™

Product Code. 10682665
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682665 5 g Amber Glass Bottle 5g
10611835 250 mg Amber Glass Bottle 250mg
10520323 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682665 Supplier Thermo Scientific Maybridge Supplier No. MO01152DE
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69682-13-9
Molecular Formula C9H20ClN
Molecular Weight (g/mol) 177.716
MDL Number MFCD05865119
InChI Key RWQQDIHTYQYXDX-UHFFFAOYSA-N
Synonym 4-tert-butyl piperidine hydrochloride, 4-tert-butylpiperidine hydrochloride, 4-tert-butyl-piperidine hydrochloride, 4-tert-butylpiperidine hcl, 4-tert-butyl piperidine, chloride
PubChem CID 2794676
IUPAC Name 4-tert-butylpiperidine;hydrochloride
SMILES CC(C)(C)C1CCNCC1.Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 177.72
Percent Purity 90%
Chemical Name or Material 4-(Tert-butyl)piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.