missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10342133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Dimensione della confezione:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
10342133 1 g 1g
10125280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10342133 Fornitore Thermo Scientific Acros N. del fornitore 308740010
 Restricted Item
Estimated Shipment: 07-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 943-27-1 | Molecular Formula: C12H16O | Molecular Weight: 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 943-27-1
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00017256
InChI Key UYFJYGWNYQCHOB-UHFFFAOYSA-N
Synonym 4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanone
PubChem CID 13669
IUPAC Name 1-(4-tert-butylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifica

CAS Min % 97.5
CAS Max % 100.0
Melting Point 17.0°C to 18.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 107.0°C to 108.0°C (5.0 mmHg)
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4COCH3
Refractive Index 1.5190 to 1.5220
Quantity 1 g
Specific Gravity 0.96
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Formula Weight 176.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4'-tert-Butylacetophenone, 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.