missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonyl chloride, 98%

Product Code. 10743341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10743341 10 g Glass bottle 10g
10762885 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743341 Supplier Thermo Scientific Acros Supplier No. 303470100
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 15084-51-2 | C10H13ClO2S | 232.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15084-51-2
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.73
MDL Number MFCD00007449
InChI Key YEZADZMMVHWFIY-UHFFFAOYSA-N
Synonym 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride
PubChem CID 139882
IUPAC Name 4-tert-butylbenzenesulfonyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 83°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4SO2Cl
Quantity 10 g
Formula Weight 232.73
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.