missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonyl Chloride, TRC

Product Code. 30345573
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345573 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345573 Supplier Toronto Research Chemicals Supplier No. TRCB8108401G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-tert-Butylbenzenesulfonyl Chloride
CAS 15084-51-2
Molecular Formula C10H13ClO2S
Quantity 1 g
Synonym 4-(1,1-Dimethylethyl)benzenesulfonyl Chloride, 4-t-Butylphenylsulfonyl Chloride, 4-tert-Butylbenzene-1-sulfonyl Chloride, 4-tert-Butylphenylsulfonyl Chloride, NSC 186263, p-(tert-Butyl)benzenesulfonyl Chloride, p-tert-Butylphenylsulfonyl Chloride, tert-Butylbenzene-4-sulfonyl Chloride
InChI Formula InChI=1S/C10H13ClO2S/c1-10(2,3)8-4-6-9(7-5-8)14(11,12)13/h4-7H,1-3H3
SMILES CC(C)(C)c1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-tert-butylbenzenesulfonyl chloride
Molecular Weight (g/mol) 232.73
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.