missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzhydrazide, 98+%

Product Code. 11440743
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440743 100 g 100g
11430743 25 g 25g
11450743 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440743 Supplier Thermo Scientific Alfa Aesar Supplier No. A13224.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 43100-38-5 | C11H16N2O | 192.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43100-38-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00014763
InChI Key XYUFQWDLRLHUPB-UHFFFAOYSA-N
Synonym 4-tert-butylbenzhydrazide, 4-tert-butyl benzohydrazide, benzoic acid, 4-1,1-dimethylethyl-, hydrazide, p-tert-butylbenzohydrazide, 4-tert-butylbenzhyrazide, 4-tert-butylbenzoic acid hydrazide, 4-tert-butylbenzoic hydrazide, 4-tert-butyl benzenecarbohydrazide, 4-tert-butyl benzene-1-carbohydrazide, 4-t-butylbenzhydrazide
PubChem CID 123513
IUPAC Name 4-tert-butylbenzohydrazide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 127°C to 131°C
Quantity 100 g
Beilstein 1104663
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material 4-tert-Butylbenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.