missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%

Product Code. 11318065
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318065 25 g 25g
11308065 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318065 Supplier Thermo Scientific Alfa Aesar Supplier No. L08248.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5451-55-8 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5451-55-8
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00042622
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
Synonym trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O

Specifications

Quantity 25 g
Beilstein 2043187
Formula Weight 184.28
Percent Purity ≥98%
Chemical Name or Material 4-tert-Butylcyclohexanecarboxylic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.