missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexene-1-boronic acid, 97%, Thermo Scientific™

Product Code. 15420038
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420038 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420038 Supplier Thermo Scientific Alfa Aesar Supplier No. H53282.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-tert-Butylcyclohexene-1-boronic acid is a versatile reagents for Suzuki-Miyaura cross-coupling. Used as protecting groups to eliminate unwanted side reactions.

Solubility
Slightly soluble in water.

Notes
TRUSTED_SUSTAINABILITY

Specifications

CAS 850567-91-8
Melting Point 166°C to 170°C
Molecular Formula C10H19BO2
MDL Number MFCD06659856
Quantity 250 mg
Synonym 4-tert-butyl cyclohex-1-en-1-yl boronic acid, 4-t-butylcyclohexen-1-ylboronic acid, 4-tert-butyl-1-cyclohexen-1-ylboronic acid, 4-tert-butylcyclohexen-1-yl boronic acid, 4-tert-butylcyclohex-1-en-1-ylboronic acid, 4-tert-butylcyclohex-1-enylboronic acid, acmc-209q0t, 4-t-butyl-cyclohex-1-enyl boronic acid, 4-t-butyl-cyclohexen-1-yl boronic acid
Solubility Information Slightly soluble in water.
InChI Key QPJWLDFYBBQYSG-UHFFFAOYSA-N
SMILES B(C1=CCC(CC1)C(C)(C)C)(O)O
IUPAC Name (4-tert-butylcyclohexen-1-yl)boronic acid
Molecular Weight (g/mol) 182.07
PubChem CID 44118553
Formula Weight 182.07
Percent Purity 97%
Chemical Name or Material 4-tert-Butylcyclohexene-1-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.