missing translation for 'onlineSavingsMsg'
Learn More

4-((tert-Butyldimethylsilyl)oxy)-2,5-bis(trifluoromethyl)aniline, TRC

Product Code. 30334353
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334353 50 mg 50mg
30329566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30334353 Supplier Toronto Research Chemicals Supplier No. TRCB94275850MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-((tert-Butyldimethylsilyl)oxy)-2,5-bis(trifluoromethyl)aniline
CAS 2512202-83-2
Molecular Formula C14 H19 F6 N O Si
Quantity 50 mg
InChI Formula InChI=1S/C14H19F6NOSi/c1-12(2,3)23(4,5)22-11-7-8(13(15,16)17)10(21)6-9(11)14(18,19)20/h6-7H,21H2,1-5H3
SMILES CC(C)(C)[Si](C)(C)Oc1cc(c(N)cc1C(F)(F)F)C(F)(F)F
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxy-2,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol) 359.383
Formula Weight 359.114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.