missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylpyridine

Product Code. 17296836 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17296836 100 g 100g
17251136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17296836 Supplier Thermo Scientific Acros Supplier No. 161941000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3978-81-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3978-81-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key YSHMQTRICHYLGF-UHFFFAOYSA-N
IUPAC Name 4-tert-butylpyridine
SMILES CC(C)(C)C1=CC=NC=C1

Specifications

Density 0.9230 g/mL
Boiling Point 196.0°C to 197.0°C
Flash Point 63°C
Linear Formula (C5H4N)C(CH3)3
Quantity 100 g
Beilstein 20, 252
Specific Gravity 0.923
Solubility Information Solubility aq. soln.: 6.9 g/l (20°C).
Formula Weight 135.22
Chemical Name or Material 4-tert-Butylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.